N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B5140-1G B5140-5G
TCIAB5140-1GEA 137 EUR
TCIAB5140-1G TCIAB5140-5G
N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine
Formule: C₄₀H₃₆N₂O₂
MDL Nummer: MFCD28384132
CAS nummer: 169685-34-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9be6e394a0903200cfbb850e0d42387bae917d9e&vid=9be6e394a0903200cfbb850e0d42387bae917d9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR