Bis(2,4-dimethylphenyl)amine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B5148-200MG B5148-1G
TCIAB5148-200MGEA 46.2 EUR
TCIAB5148-200MG TCIAB5148-1G
Bis(2,4-dimethylphenyl)amine ≥98.0% (by GC)
Bis(2,4-dimethylphenyl)amine
Formule: C₁₆H₁₉N
Kookpunt: 310°C
MDL Nummer: MFCD16619146
CAS nummer: 19616-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a8e35f3976ae2b1024c00e917c402d70e5a89b5&vid=5a8e35f3976ae2b1024c00e917c402d70e5a89b5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C