4-Bromo-3-iodotoluene ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B5168-1G B5168-5G
TCIAB5168-1GEA 57.2 EUR
TCIAB5168-1G TCIAB5168-5G
4-Bromo-3-iodotoluene ≥96.0% (by GC)
4-Bromo-3-iodotoluene
Formule: C₇H₆BrI
Kookpunt: 150°C
MDL Nummer: MFCD07784420
CAS nummer: 858841-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dde723748252d35bb47f9c24896a247e98af664&vid=0dde723748252d35bb47f9c24896a247e98af664&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR