(3-Bromophenyl)triphenylsilane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B5222-200MG B5222-1G
TCIAB5222-200MGEA 27.5 EUR
TCIAB5222-200MG TCIAB5222-1G
(3-Bromophenyl)triphenylsilane ≥98.0% (by GC)
(3-Bromophenyl)triphenylsilane
Formule: C₂₄H₁₉BrSi
MDL Nummer: MFCD28098175
CAS nummer: 185626-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fadc24791e34c5d500225b6d8fdf2e7d0a9e2137&vid=fadc24791e34c5d500225b6d8fdf2e7d0a9e2137&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 135.0 to 139.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR