2-Bromo-3-fluorophenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B5224-1G B5224-5G
TCIAB5224-1GEA 38.5 EUR
TCIAB5224-1G TCIAB5224-5G
2-Bromo-3-fluorophenol ≥98.0% (by GC)
2-Bromo-3-fluorophenol
Formule: C₆H₄BrFO
Kookpunt: 123°C
MDL Nummer: MFCD04037220
CAS nummer: 443-81-2
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %