4'-Bromo-2'-fluoroacetophenone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B5236-1G B5236-5G
TCIAB5236-1GEA 40.7 EUR
TCIAB5236-1G TCIAB5236-5G
4'-Bromo-2'-fluoroacetophenone ≥98.0% (by GC)
4'-Bromo-2'-fluoroacetophenone
Formule: C₈H₆BrFO
MDL Nummer: MFCD03411551
CAS nummer: 625446-22-2
Order Now
Specification Test Results
Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %