Benzyl anthranilate ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B5249-1G B5249-5G
TCIAB5249-1GEA 29.7 EUR
TCIAB5249-1G TCIAB5249-5G
Benzyl anthranilate ≥98.0% (by GC, titration analysis)
Benzyl anthranilate
Formule: C₁₄H₁₃NO₂
MW: 227.26 g/mol
MDL Nummer: MFCD00060605
CAS nummer: 82185-41-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e8ec9e53640bc04a92e6e2b3ecff195799f0ac2&vid=5e8ec9e53640bc04a92e6e2b3ecff195799f0ac2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C