2-Bromo-4-(tert-butyl)aniline ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B5325-25G
B5325-5G
TCIAB5325-25GEA
195
EUR
TCIAB5325-25G
TCIAB5325-5G
2-Bromo-4-(tert-butyl)aniline ≥98.0% (by GC, titration analysis)
2-Bromo-4-(tert-butyl)aniline
Formule:
C₁₀H₁₄BrN Kookpunt: 265°C |
MDL Nummer:
MFCD00052693 CAS nummer: 103273-01-4 |
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...