1-Bromo-4-fluorobutane ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B5331-5G
B5331-25G
TCIAB5331-5GEA
52.8
EUR
TCIAB5331-5G
TCIAB5331-25G
1-Bromo-4-fluorobutane ≥98.0% (by GC)
1-Bromo-4-fluorobutane
Formule:
BrH₂C(CH₂)₂CH₂F Kookpunt: 135°C Vlampunt: 45°C |
MDL Nummer:
MFCD00209551 CAS nummer: 462-72-6 UN: 1993 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...