(2,4-Bis(trifluoromethyl)phenyl)acetic acid ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2,4-Bis(trifluoromethyl)phenylacetic acid

B5337-1G B5337-5G
TCIAB5337-1GEA 83.6 EUR
TCIAB5337-1G TCIAB5337-5G
(2,4-Bis(trifluoromethyl)phenyl)acetic acid ≥97.0% (by titrimetric analysis)
(2,4-Bis(trifluoromethyl)phenyl)acetic acid
Formule: (C₆H₃(CF₃)₂)OCH₂COOH
MDL Nummer: MFCD00042492
CAS nummer: 177952-39-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR