(tert-Butyldimethylsilyloxy)malononitrile ≥93.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B5370-200MG B5370-1G
TCIAB5370-200MGEA 131 EUR
TCIAB5370-200MG TCIAB5370-1G
(tert-Butyldimethylsilyloxy)malononitrile ≥93.0% (by GC)
(tert-Butyldimethylsilyloxy)malononitrile
Formule: C₉H₁₆N₂OSi
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD27942977
CAS nummer: 128302-78-3
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f43b516e62cf70386e7d15b69e6f0bebfb51458&vid=3f43b516e62cf70386e7d15b69e6f0bebfb51458&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR