4-tert-Butylcalix[6]arene
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: p-tert-Butylcalix[6]arene
B5375-1G B5375-5G
TCIAB5375-1GEA 20.9 EUR
TCIAB5375-1G TCIAB5375-5G
4-tert-Butylcalix[6]arene
4-tert-Butylcalix[6]arene
Formule: C₆₆H₈₄O₆
MW: 973.39 g/mol
Smeltpunt: 381°C
MDL Nummer: MFCD00075464
CAS nummer: 78092-53-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9389afd0f7f1b3c66a393bed1c85913d31b2707&vid=f9389afd0f7f1b3c66a393bed1c85913d31b2707&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area% (Except for the toluene)
Residual Solvent max. 10.0 %