4'-Bromo-2'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1-(4-Bromo-2-hydroxyphenyl)ethanone, 1-(4-Bromo-2-hydroxyphenyl)ethan-1-one

B5382-1G B5382-5G
TCIAB5382-1GEA 27.5 EUR
TCIAB5382-1G TCIAB5382-5G
4'-Bromo-2'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
4'-Bromo-2'-hydroxyacetophenone
Formule: C₈H₇BrO₂
MW: 215.05
Kookpunt: 83°C
MDL Nummer: MFCD03428533
CAS nummer: 30186-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=642b75dfa90f870daa2315c703d12f47ac774e2c&vid=642b75dfa90f870daa2315c703d12f47ac774e2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR