2-Bromo-5-dodecylthiophene ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B5407-1G B5407-5G
TCIAB5407-1GEA 80.3 EUR
TCIAB5407-1G TCIAB5407-5G
2-Bromo-5-dodecylthiophene ≥95.0% (by GC)
2-Bromo-5-dodecylthiophene
Formule: C₁₆H₂₇BrS
Kookpunt: 162°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD22572689
CAS nummer: 153561-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=717b5d66558e0c9793ee13f20855099172f7d1bb&vid=717b5d66558e0c9793ee13f20855099172f7d1bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green lump
Purity (GC) min. 95.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR