2-Bromo-3-tetradecylthiophene ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B5408-1G B5408-5G
TCIAB5408-1GEA 158 EUR
TCIAB5408-1G TCIAB5408-5G
2-Bromo-3-tetradecylthiophene ≥95.0% (by GC)
2-Bromo-3-tetradecylthiophene
Formule: C₁₈H₃₁BrS
CAS nummer: 500199-09-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76051d9ad65e1467fd16c566e99055bbd021e944&vid=76051d9ad65e1467fd16c566e99055bbd021e944&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR