4-Bromophthalonitrile ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 4-Bromo-1,2-dicyanobenzene

B5509-1G B5509-5G
TCIAB5509-1GEA 64.9 EUR
TCIAB5509-1G TCIAB5509-5G
4-Bromophthalonitrile ≥97.0% (by GC)
4-Bromophthalonitrile
Formule: C₈H₃BrN₂
MDL Nummer: MFCD00221794
CAS nummer: 70484-01-4
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd52beb5ed6cc200a322078ea297f1b1d27ea809&vid=cd52beb5ed6cc200a322078ea297f1b1d27ea809&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 138.0 to 142.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR