1-Butyl-3-methylimidazoliumtrifluoroacetate ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B5526-5G B5526-25G
TCIAB5526-5GEA 79.2 EUR
TCIAB5526-5G TCIAB5526-25G
1-Butyl-3-methylimidazoliumtrifluoroacetate ≥97.0% (by titrimetric analysis)
1-Butyl-3-methylimidazoliumtrifluoroacetate
Formule: C₁₀H₁₅F₃N₂O₂
MDL Nummer: MFCD08458921
CAS nummer: 174899-94-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=552c3b6f054fe6f8f0f950368afd68feb854148e&vid=552c3b6f054fe6f8f0f950368afd68feb854148e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR