2-Butyl-n-octan-1-amine ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B5550-1ML
B5550-5ML
TCIAB5550-1MLEA
121
EUR
TCIAB5550-1ML
TCIAB5550-5ML
2-Butyl-n-octan-1-amine ≥98.0% (by titrimetric analysis)
2-Butyl-n-octan-1-amine
Formule:
C₁₂H₂₇N |
MDL Nummer:
MFCD20721903 CAS nummer: 217655-07-7 UN: 2735 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...