2-tert-Butoxypyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B5575-1G B5575-5G
TCIAB5575-1GEA 121 EUR
TCIAB5575-1G TCIAB5575-5G
2-tert-Butoxypyridine ≥98.0% (by GC)
2-tert-Butoxypyridine
Formule: C₉H₁₃NO
Kookpunt: 88°C
MDL Nummer: MFCD00234567
CAS nummer: 83766-88-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR