3-Bromobenzo[b]thiophene ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B5589-5G B5589-25G
TCIAB5589-5GEA 53.9 EUR
TCIAB5589-5G TCIAB5589-25G
3-Bromobenzo[b]thiophene ≥96.0% (by GC)
3-Bromobenzo[b]thiophene
Formule: C₈H₅BrS
Kookpunt: 269°C
MDL Nummer: MFCD00023009
CAS nummer: 7342-82-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a9b10acb83cf581cd68ef49ab5e55a6238f7ed8&vid=3a9b10acb83cf581cd68ef49ab5e55a6238f7ed8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 96.0 %