Benztropine mesylate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B5592-5G B5592-25G
TCIAB5592-5GEA 84.7 EUR
TCIAB5592-5G TCIAB5592-25G
Benztropine mesylate ≥98.0% (by HPLC)
Benztropine mesylate
Formule: C₂₂H₂₉NO₄S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00074784
CAS nummer: 132-17-2
EINECS: 205-048-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,01121

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR