1,3-Bis(methoxymethoxy)benzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Resorcinol bis(methoxymethyl) ether, 1,3-Di(methoxymethoxy)benzene
B5643-1G B5643-5G
TCIAB5643-1GEA 53.9 EUR
TCIAB5643-1G TCIAB5643-5G
1,3-Bis(methoxymethoxy)benzene ≥98.0% (by GC)
1,3-Bis(methoxymethoxy)benzene
Formule: C₁₀H₁₄O₄
MW: 198.22 g/mol
Kookpunt: 137°C
MDL Nummer: MFCD27925713
CAS nummer: 57234-29-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f05eb0fcb4866cce54985c04ffcb9ba8ae251a7&vid=4f05eb0fcb4866cce54985c04ffcb9ba8ae251a7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %