3-Bromopropyl Acetate ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B5650-1G
B5650-5G
TCIAB5650-1GEA
56.1
EUR
TCIAB5650-1G
TCIAB5650-5G
3-Bromopropyl Acetate ≥95.0% (by GC)
3-Bromopropyl Acetate
Formule:
C₅H₉BrO₂ Kookpunt: 163°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD02258489 CAS nummer: 592-33-6 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...