1-Bromo-2,4-difluoro-5-nitrobenzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 5-Bromo-2,4-difluoronitrobenzene

B5688-1G B5688-5G
TCIAB5688-1GEA 19.8 EUR
TCIAB5688-1G TCIAB5688-5G
1-Bromo-2,4-difluoro-5-nitrobenzene ≥98.0% (by GC)
1-Bromo-2,4-difluoro-5-nitrobenzene
Formule: C₆H₂BrF₂NO₂
Kookpunt: 97°C
Smeltpunt: 19°C
MDL Nummer: MFCD00129166
CAS nummer: 345-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=818b3562a52b00e9f35d32469e9e559997096e91&vid=818b3562a52b00e9f35d32469e9e559997096e91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR