3-Benzyloxybenzyl bromide ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 3-(Benzyloxy)benzyl bromide, 1-(Benzyloxy)-3-(bromomethyl)benzene

B5708-1G B5708-5G
TCIAB5708-1GEA 86.9 EUR
TCIAB5708-1G TCIAB5708-5G
3-Benzyloxybenzyl bromide ≥95.0% (by GC)
3-Benzyloxybenzyl bromide
Formule: C₁₄H₁₃BrO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01318307
CAS nummer: 1700-31-8
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR