4-(Pyrimidin-2-yl)bromobenzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B5806-1G B5806-200MG
TCIAB5806-1GEA 237 EUR
TCIAB5806-1G TCIAB5806-200MG
4-(Pyrimidin-2-yl)bromobenzene ≥98.0% (by GC)
4-(Pyrimidin-2-yl)bromobenzene
Formule: C₁₀H₇BrN₂
MDL Nummer: MFCD08056329
CAS nummer: 210354-17-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR