2-(3-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Stabilisatie: Stabilised with phenothiazine

B5960-1G B5960-5G
TCIAB5960-1GEA 82.5 EUR
TCIAB5960-1G TCIAB5960-5G
2-(3-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC) stabilised
2-(3-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formule: C₁₀H₁₉BO₂
MDL Nummer: MFCD11858602
CAS nummer: 331958-92-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR