1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B5985-100MG
B5985-500MG
TCIAB5985-100MGEA
236
EUR
TCIAB5985-100MG
TCIAB5985-500MG
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane ≥95.0% (by GC)
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane
Formule:
C₁₄H₂₈P₂ Kookpunt: 67°C |
MDL Nummer:
MFCD01073770 CAS nummer: 129648-07-3 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...