4'-Benzyloxy-2-bromopropiophenone ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B5990-5G
B5990-25G
TCIAB5990-5GEA
38.5
EUR
TCIAB5990-5G
TCIAB5990-25G
4'-Benzyloxy-2-bromopropiophenone ≥98.0% (by GC)
4'-Benzyloxy-2-bromopropiophenone
Formule:
C₁₆H₁₅BrO₂ |
MDL Nummer:
MFCD00017862 CAS nummer: 35081-45-9 EINECS: 252-351-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...