1-Bromo-5-fluoropentane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B5998-5G B5998-25G
TCIAB5998-5GEA 20.9 EUR
TCIAB5998-5G TCIAB5998-25G
1-Bromo-5-fluoropentane ≥98.0% (by GC)
1-Bromo-5-fluoropentane
Formule: C₅H₁₀BrF
Kookpunt: 162°C
MDL Nummer: MFCD01709395
CAS nummer: 407-97-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR