2-Butylbenzo[d]isothiazol-3(2H)-one ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B6021-25G B6021-100G
TCIAB6021-25GEA 68.2 EUR
TCIAB6021-25G TCIAB6021-100G
2-Butylbenzo[d]isothiazol-3(2H)-one ≥95.0% (by GC)
2-Butylbenzo[d]isothiazol-3(2H)-one
Formule: C₁₁H₁₃NOS
Kookpunt: 180°C
MDL Nummer: MFCD09751282
CAS nummer: 4299-07-4
EINECS: 420-590-7
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR