2-tert-Butyl-1,1,3,3-tetramethylguanidine ≥95.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 2-(tert-Butyl)-1,1,3,3-tetramethylguanidine, BTMG

B6052-1G B6052-5G
TCIAB6052-1GEA 37.4 EUR
TCIAB6052-1G TCIAB6052-5G
2-tert-Butyl-1,1,3,3-tetramethylguanidine ≥95.0% (by GC, titration analysis)
2-tert-Butyl-1,1,3,3-tetramethylguanidine
Formule: C₉H₂₁N₃
MW: 171.29 g/mol
Vlampunt: 65°C
MDL Nummer: MFCD01862972
CAS nummer: 29166-72-1
UN: 3267
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR