Benzyl Benzodithioate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Benzyl benzodithioate, Benzyl Benzenecarbodithioate, Benzodithioic acid Benzyl ester, Benzenecarbodithioic acid Benzyl ester
B6066-1G B6066-5G
TCIAB6066-1GEA 160 EUR
TCIAB6066-1G TCIAB6066-5G
Benzyl Benzodithioate ≥95.0% (by GC)
Benzyl Benzodithioate
Formule: C₁₄H₁₂S₂
MW: 244.37 g/mol
Kookpunt: 254°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD08705657
CAS nummer: 27249-90-7
Order Now
Specification Test Results
Appearance Orange to Brown to Dark red clear liquid
Purity (GC) min. 95.0 %