Boc-L-β-homophenylalanine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Boc-L-beta-Homophe-OH, (S)-3-[(tert-Butoxycarbonyl)amino]-4-phenylbutanoic acid, Boc-β-HoPhe-OH, Boc-β-HomoPhe-OH, (S)-3-[(tert-Butoxycarbonyl)amino]-4-phenylbutyric acid, Boc-L-beta-homophenylalanine, N-(tert-Butoxycarbonyl)-L-beta-homophenylalanine

B6074-250MG B6074-1G
TCIAB6074-250MGEA 52.8 EUR
TCIAB6074-250MG TCIAB6074-1G
Boc-L-β-homophenylalanine ≥98.0% (by HPLC, titration analysis)
Boc-L-beta-homophenylalanine
Formule: C₁₅H₂₁NO₄
MDL Nummer: MFCD01076271
CAS nummer: 51871-62-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR