Boc-L-cyclopropylglycine

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B6088-1G B6088-200MG
TCIAB6088-1GEA 143 EUR
TCIAB6088-1G TCIAB6088-200MG
Boc-L-cyclopropylglycine
Boc-L-cyclopropylglycine
Formule: C₁₀H₁₇NO₄
Smeltpunt: 112°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06659132
CAS nummer: 155976-13-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Melting point 116.0 to 120.0 deg-C
Specific rotation [a]20/D +35.0 to +41.0 deg (C=1, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR