1-Benzyl-5-bromoindole ≥98.0% (by GC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-Benzyl-5-bromo-1H-indole

B6090-1G B6090-5G
TCIAB6090-1GEA 60.5 EUR
TCIAB6090-1G TCIAB6090-5G
1-Benzyl-5-bromoindole ≥98.0% (by GC, total nitrogen)
1-Benzyl-5-bromoindole
Formule: C₁₅H₁₂BrN
MW: 286.17 g/mol
MDL Nummer: MFCD04337704
CAS nummer: 10075-51-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR