Butyl 2-Methylbutyrate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B6101-100ML B6101-25ML
TCIAB6101-100MLEA 74.8 EUR
TCIAB6101-100ML TCIAB6101-25ML
Butyl 2-Methylbutyrate ≥98.0% (by GC)
Butyl 2-Methylbutyrate
Formule: C₉H₁₈O₂
Kookpunt: 176°C
Vlampunt: 60°C
MDL Nummer: MFCD00042902
CAS nummer: 15706-73-7
EINECS: 239-798-2
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %