Guaiacol carbonate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Carbonic Acid Bis(2-methoxyphenyl) Ester, Bis(2-methoxyphenyl) Carbonate

C0039-25G C0039-500G
TCIAC0039-25GEA 61.6 EUR
TCIAC0039-25G TCIAC0039-500G
Guaiacol carbonate ≥98.0% (by HPLC)
Guaiacol carbonate
Formule: C₁₅H₁₄O₅
MW: 274.27 g/mol
MDL Nummer: MFCD00025723
CAS nummer: 553-17-3
EINECS: 209-034-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 87.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR