Allyl chloroacetate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Chloroacetic acid allyl ester, Allyl 2-chloroacetate

C0090-25G C0090-500G
TCIAC0090-25GEA 40.7 EUR
TCIAC0090-25G TCIAC0090-500G
Allyl chloroacetate ≥98.0% (by GC)
Allyl chloroacetate
Formule: C₅H₇ClO₂
Kookpunt: 164°C
Vlampunt: 61°C
MDL Nummer: MFCD00058936
CAS nummer: 2916-14-5
EINECS: 220-839-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR