4-Chloroaniline-3-sulphonic acid ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
5-Amino-2-chlorobenzenesulphonic acid
C0116-25G
C0116-500G
TCIAC0116-25GEA
20.9
EUR
TCIAC0116-25G
TCIAC0116-500G
4-Chloroaniline-3-sulphonic acid ≥98.0% (by HPLC, titration analysis)
4-Chloroaniline-3-sulphonic acid
Formule:
C₆H₆ClNO₃S MW: 207.63 g/mol |
MDL Nummer:
MFCD00025296 CAS nummer: 88-43-7 EINECS: 201-830-8 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...