Isobutyl chloroformate ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Chloroformic acid isobutyl ester
C0178-25G
C0178-100G
C0178-500G
TCIAC0178-25GEA
18.7
EUR
TCIAC0178-25G
TCIAC0178-100G
TCIAC0178-500G
Isobutyl chloroformate ≥98.0% (by titrimetric analysis)
Isobutyl chloroformate
Formule:
C₅H₉ClO₂ MW: 136.58 g/mol Kookpunt: 129°C Smeltpunt: -80 Vlampunt: 35°C |
MDL Nummer:
MFCD00000642 CAS nummer: 543-27-1 EINECS: 208-840-1 UN: 2742 ADR: 6.1 II Merck Index: 14,05139 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...