5-Chloro-2-hydroxybenzophenone ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
(5-Chloro-2-hydroxyphenyl)(phenyl)methanone
C0186-5G
C0186-25G
TCIAC0186-5GEA
64.9
EUR
TCIAC0186-5G
TCIAC0186-25G
5-Chloro-2-hydroxybenzophenone ≥98.0% (by GC)
5-Chloro-2-hydroxybenzophenone
Formule:
C₁₃H₉ClO₂ MW: 232.6600 g/mol |
MDL Nummer:
MFCD00020134 CAS nummer: 85-19-8 EINECS: 201-592-5 |
Specification Test Results
Purity (GC) | min. 98.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...