4-Chlorophenylhydrazine sulphate ≥95.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C0258-25G
C0258-500G
TCIAC0258-25GEA
51.7
EUR
TCIAC0258-25G
TCIAC0258-500G
4-Chlorophenylhydrazine sulphate ≥95.0% (by titrimetric analysis)
4-Chlorophenylhydrazine sulphate
Formule:
C₆H₇ClN₂ |
MDL Nummer:
MFCD00043829 CAS nummer: 14581-21-6 EINECS: 238-624-2 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Potassium iodate Method) | min. 95.0 % |
Solubility in hot Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...