Chondroitin sulphate sodium salt
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C0335-25G
C0335-100G
TCIAC0335-25GEA
102
EUR
TCIAC0335-25G
TCIAC0335-100G
Chondroitin sulphate sodium salt
Chondroitin sulphate sodium salt
MDL Nummer:
MFCD00130792 CAS nummer: 9082-07-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
pH | 5.5 to 7.5 (10g/L, 25 deg-C) |
Drying loss | max. 10.0 % |
Ignition residue (Sulfate) | 23.0 to 31.0 % (calcd.on dried substance) |
Total Nitrogen | 2.5 to 3.8 % (calcd.on dried substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...