Croton oil
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Tiglium oil
C0421-25ML
C0421-5ML
TCIAC0421-25MLEA
173
EUR
TCIAC0421-25ML
TCIAC0421-5ML
Croton oil
Croton oil
MDL Nummer:
MFCD00130880 CAS nummer: 8001-28-3 UN: 2810 ADR: 6.1 III Merck Index: 14,02598 |
Specification Test Results
Appearance | Orange to Amber to Dark red clear liquid |
Iodine Value | 100 to 120 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...