Cyanine ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
1-Isopentyl-4-((1-isopentylquinolin-4(1H)-ylidene)methyl)quinolin-1-ium iodide, Quinoline Blue
C0436-1G
C0436-5G
TCIAC0436-1GEA
104
EUR
TCIAC0436-1G
TCIAC0436-5G
Cyanine ≥98.0% (by titrimetric analysis)
Cyanine
Formule:
C₂₉H₃₅IN₂ |
MDL Nummer:
MFCD00050330 CAS nummer: 523-42-2 EINECS: 208-345-0 |
Specification Test Results
Appearance | Green to Dark green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on dried substance) |
Lambda max. | 591.0 to 594.0 nm (MeOH) |
Molar extinction coefficient | min. 85000 (MeOH、591.0 to 594.0 nm) |
Drying loss | max. 3.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...