Cytidine hemisulphate salt ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Cytidine sulphate, Cytosine β-D-riboside

C0525-1G C0525-100MG
TCIAC0525-1GEA 33 EUR
TCIAC0525-1G TCIAC0525-100MG
Cytidine hemisulphate salt ≥98.0% (by HPLC, titration analysis)
Cytidine hemisulphate salt
Formule: C₁₈H₂₆N₆O₁₀·H₂SO₄
MDL Nummer: MFCD00151091
CAS nummer: 32747-18-5
EINECS: 251-191-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +35.0 to +38.0 deg (C=2, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR