Isopropyl carbamate ≥98.0% (by GC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Carbamic Acid Isopropyl Ester

C0614-25G C0614-250G
TCIAC0614-25GEA 72.6 EUR
TCIAC0614-25G TCIAC0614-250G
Isopropyl carbamate ≥98.0% (by GC, total nitrogen)
Isopropyl carbamate
Formule: C₄H₉NO₂
Kookpunt: 183°C
MDL Nummer: MFCD00025463
CAS nummer: 1746-77-6
EINECS: 217-127-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 92.0 to 95.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR