N-Carbobenzoxy-L-proline ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Z-Pro-OH, Z-L-Prolin, N-Cbz-L-proline, N-Carbobenzyloxy-L-proline, N-Benzyloxycarbonyl-L-proline

C0713-25G C0713-5G
TCIAC0713-25GEA 53.9 EUR
TCIAC0713-25G TCIAC0713-5G
N-Carbobenzoxy-L-proline ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-L-proline
Formule: C₁₃H₁₅NO₄
MW: 249.27 g/mol
MDL Nummer: MFCD00003170
CAS nummer: 1148-11-4
EINECS: 214-557-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 76.0 to 78.0 deg-C
Specific rotation [a]20/D -38.0 to -42.0 deg (C=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR