Methyl-3-cyanopropionate ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C0792-25ML C0792-5ML
TCIAC0792-25MLEA 484 EUR
TCIAC0792-25ML TCIAC0792-5ML
Methyl-3-cyanopropionate ≥99.0% (by GC)
Methyl-3-cyanopropionate
Formule: C₅H₇NO₂
Kookpunt: 216°C
MDL Nummer: MFCD00058965
CAS nummer: 4107-62-4
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR